Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Improves laboratory workflow
Brand:  SGE™ 2910020
 View more versions of this product
Product Code. 12927586
  • 104.00 € / Each
Explore more special offers
Description
Description
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
  • Digitally controlled positive displacement dispensing system
  • Programmable—enables user-independent liquid handling procedures
  • Easily calibrated to ensure accurate dispensing
  • Stores calibration factors for up to ten XCHANGE™ syringes
  • Touch Wheel user interface
  • Full color screen
Applications:
  • Preparation of calibration standards
  • Addition of internal standards
  • Precise dispensing of non-aqueous liquids
  • Sample dilution
Specifications
Specifications
25 G
0.12 mm
0.5 mm
1.96 in.
Ea.
Bevel
0.0047 in.
0.019 in.
Preparation of calibration standards, addition of internal standards
eVol Analytical Syringe
5 μL
eVol Automated Syringe