Learn More
SGE™ eVol™ Handheld Automated Analytical Syringe
Improves laboratory workflow
Brand: SGE™ 2910022
This item is not returnable.
View return policy
Description
eVol™ is the coupling of two precision devices—a digitally controlled electronic drive and an XCHANGE™ (patent pending) enabled analytical syringe. The XCHANGE™ analytical syringes can be easily and quickly changed, allowing them to be dedicated to individual liquids and methods, while preventing cross-contamination of reagents.
- Digitally controlled positive displacement dispensing system
- Programmable—enables user-independent liquid handling procedures
- Easily calibrated to ensure accurate dispensing
- Stores calibration factors for up to ten XCHANGE™ syringes
- Touch Wheel user interface
- Full color screen
Applications:
- Preparation of calibration standards
- Addition of internal standards
- Precise dispensing of non-aqueous liquids
- Sample dilution
Specifications
25 G | |
0.2 mm | |
0.5 mm | |
1.96 in. | |
Ea. | |
Bevel |
0.007 in. | |
0.019 in. | |
Preparation of calibration standards, addition of internal standards | |
eVol Analytical 25 ga | |
50 μL | |
eVol Automated Syringe |