Learn More
Trajan™ eVol™ MEPS™ Syringes
For use with eVol™ Digital Analytical Syringe digitally controlled positive displacement dispensing system.
Brand:  Trajan™ 2910028
Product Code. 12987586
  • 133.00 € / Each
Explore more special offers
Description
Description
  • Enables MEPS (micro SPE) to be semi-automated and proofed before transition to fully automated platforms
  • 2011 Firmware required
  • Maximum operating temperature of 350°C
Specifications
Specifications
Liquid handling procedure
eVol™ Syringe
MEPS BINs
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Trajan™ eVol™ MEPS™ Syringes > eVol MEPS Syringe; 100uL