Learn More
Trajan™ eVol™ XCHANGE Syringes
XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.
Brand:  Trajan™ 2910035
 View more versions of this product
Product Code. 13467878
  • 104.00 € / Each
Explore more special offers
Description
Description
  • Liquid handling user independent device
  • Reduction in need for re-analysis
  • Syringes easily and quickly changed
  • Replacement parts available
Specifications
Specifications
23 G
1.9 in.
eVol™ Syringe
1 mL
Bevel
Liquid handling procedure
50 mm
1 Pk.
50 mm