Learn More
SGE™ BP20 (Wax) GC Capillary Columns
Combines innovative phase technology and unique fused silica production capability. SGE™ BP20 (Wax) GC Capillary Columns provides end-to-end separation solutions for all applications.
Brand:  SGE™ 054437
 View more versions of this product
Product Code. 12985581
  • 1340.00 € / Each
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Features
  • Industry standard Wax column
  • Bonded and cross-linked
  • Can be solvent rinsed
Applications
  • Used for separation of Alcohol, Free acids, Fatty acids methyl esters, Aromatics, Solvents and Essential oils
Specifications
Specifications
280°C
0.32 mm
G14, G15, G16, G20, G39
20°C to 260/280°C
50 m
Fused Silica
Hydrogen Bonding or Dipole interactions
SGE™ BP20 (Wax)
Polynuclear Aromatic Hydrocarbons Analysis, Aldehydes Analysis, Ketones, PCB Congener Analysis, Nitro-substituted Aromatics, Polynuclear Aromatic Hydrocarbons, Pesticides and Herbicides Analysis
0.5 μm
Polar