missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGE™ Gas Tight Plunger Syringe with PTFE Luer Lock
Headspace syringes with a special Luer lock probe
Brand: SGE™ 008975
This item is not returnable.
View return policy
Description
- For soil gas sampling
- Special Luer lock probe has a side hole for efficient sample intake
- Fitted with a valve (SVLL)
- Gas tight
- Volume: 10mL
Specifications
For soil gas sampling | |
Plunger Syringe | |
10 mL |
PTFE luer-lock valve | |
Ea. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction