Learn More
SGE™ MEPS™ Syringes
For use in micro-SPE
Brand:  SGE™ 2910026
 View more versions of this product
Product Code. 12917596
  • 133.00 € / Each
Explore more special offers
Description
Description
  • The eVol™ custom programming function enables MEPS to be semi-automated and proofed before transition to fully automated platforms
Specifications
Specifications
Gas/Liquid chromatography
Ea.
eVol Automated Syringe
Syringe
500 μL