Learn More
SGE™ NanoVolume Syringes
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
Brand:  SGE™ 000852
 View more versions of this product
Product Code. 10578302
  • 168.00 € / Each
Explore more special offers
Description
Description
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
  • Ability to inject down to 50nL with high precision and accuracy
  • Ideal for dispensing very small liquid volumes
  • Plunger extends into the needle tip
  • Sample is contained only in the needle without any glass contact
  • Virtually zero dead volume and carry over between injections
  • Recommended for use when sample volume is less than 1000nL (1μL)
Specifications
Specifications
±2%
23 G
0.315 in.
0.63 mm
2.75 in.
5BR-7-RAX
5 μL
Borosilicate Glass (Barrel)
0.0248 in.
8 mm
Repeating adaptor
NanoVolume Syringe
Ea.
Cone