Learn More
SGE™ NanoVolume Syringes
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
Brand:  SGE™ 000803
 View more versions of this product
Product Code. 11902595
  • 117.00 € / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Ideal for nanovolume capillary chromatography injection as well as preparing accurate standards which require small volumes.
  • Ability to inject down to 50nL with high precision and accuracy
  • Ideal for dispensing very small liquid volumes
  • Plunger extends into the needle tip
  • Sample is contained only in the needle without any glass contact
  • Virtually zero dead volume and carry over between injections
  • Recommended for use when sample volume is less than 1000nL (1μL)
Specifications
Specifications
±2%
23 G
0.315 in.
0.63 mm
2.75 in.
NanoVolume Syringe
Ea.
Removable Bevel
Borosilicate Glass (Barrel)
0.0248 in.
8 mm
2.75 in.
70 mm
5BR-7BV
5 μL
Removable