Learn More
Trajan™ PerkinElmer Syringe
Equipped with cone-tipped needle.
Brand:  Trajan™ 000475
Product Code. 10299050
  • 181.00 € / Each
Explore more special offers
Description
Description
  • Specifically designed for PerkinElmer instruments
Specifications
Specifications
26 G
0.47 mm
Beveled Cone
0.155 mm
0.5 μL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products