Learn More
Trajan™ Replacement Needles for Luer-Lock Syringes
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Brand:  Trajan™ 039802
 View more versions of this product
Product Code. 11951144
  • 66.35 € / Pack of 5
Explore more special offers
Description
Description
Designed for optimum septum penetration and prevention of septum coring.
  • Standard general purpose needle
  • Made of stainless steel
  • Preferred for manual injection where piercing the septum in exactly the same place is difficult
Specifications
Specifications
Stainless Steel
0.012 in.
0.32 mm
Gas chromatography
Replacement Needle
Luer Lock
23 G
0.024 in.
0.63 mm
2 in.
Bevel