Learn More
Trajan™ Replacement Needles for Luer-Lock Syringes
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Brand: Trajan™ 039842
This item is not returnable.
View return policy
Description
Designed for optimum septum penetration and prevention of septum coring.
- Standard general purpose needle
- Made of stainless steel
- Preferred for manual injection where piercing the septum in exactly the same place is difficult
Specifications
Stainless Steel | |
0.031 in. | |
0.8 mm | |
Gas chromatography | |
Replacement Needle | |
Luer Lock |
18 G | |
0.05 in. | |
1.27 mm | |
2 in. | |
Bevel |