Learn More
Trajan™ Replacement Needles for Luer-Lock Syringes
Bevel tip style. Trajan Scientific and Medical™ Replacement Needles for Luer-Lock Syringes are supplied in packs of 5.
Brand:  Trajan™ 039807
 View more versions of this product
Product Code. 11989427
  • 67.65 € / Pack of 5
Explore more special offers
Description
Description
Designed for optimum septum penetration and prevention of septum coring.
  • Standard general purpose needle
  • Made of stainless steel
  • Preferred for manual injection where piercing the septum in exactly the same place is difficult
Specifications
Specifications
Stainless Steel
0.012 in.
0.32 mm
Gas chromatography
Replacement Needle
68.58 mm
Luer Lock
23 G
0.024 in.
0.63 mm
2.7 in.
Trajan
Bevel