missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ Fixed Needle Syringe for Beckman/Altex, Rheodyne, SSI, Valco Valves

Fixed needle replacement for manual syringes. For use with Beckman/Altex, Rheodyne, SSI, and Valco Valves

57.85 € - 87.05 €

Specifications

Material Borosilicate Glass (Barrel)
Needle Gauge 22 G
Diameter (Metric) Inner Needle 0.37 mm
Diameter (Metric) Outer Needle 0.028 mm
Product Type Manual Syringe
View More Specs

Products 4
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10462721
View Documents
Trajan™
004300
50 μL
60.30 €
Each
Please to purchase this item. Need a web account? Register with us today!
10627171
View Documents
Trajan™
003300
25 μL
57.85 €
Each
Please to purchase this item. Need a web account? Register with us today!
10025480
View Documents
Trajan™
007300
500 μL
87.05 €
Each
Please to purchase this item. Need a web account? Register with us today!
10209060
View Documents
Trajan™
006300
250 μL
74.45 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • High quality solutions for liquid handling
  • Accurate and reproducible injections
  • Specifically designed for Beckman/Altex, Rheodyne, Valco, SSI valves
Specifications

Specifications

Borosilicate Glass (Barrel)
0.37 mm
Manual Syringe
Beckman/Altex, Rheodyne, SSI, Valco Valves
22 G
0.028 mm
LC
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.