Learn More
Trajan™ Replacement Plungers
Replacement Plungers are available for all SGE gas tight syringes.
Brand: Trajan™ 031811
This item is not returnable.
View return policy
Description
- For use with SGE gas tight syringes.
- Use the SGE Syringe Part Number to find a replacement plunger.
Specifications
PTFE (Tip) | |
IN VITRO use only. For lab analyzers only. Not for human use. | |
Replacement Plunger, P10R-GT | |
Removable Needle Gas Tight Syringe (10μL) |