Learn More
Trajan™ SI-7 Solids Syringe
For use with Pyrojector to thermally crack non-volatile molecules into volatile fragements for analysis by GC.
Brand:  Trajan™ 009980
Product Code. 15298513
  • 155.00 € / Each
Explore more special offers
Description
Description
  • For use with Pyrolysis chromatography applications
  • Works in conjunction with Pyrojector II which thermally fragments essentially non-volatile molecules into volatile fragments, making them suitable for chromatographic analysis
Specifications
Specifications
Stainless Steel
For solids injection on SGE pyrojector
2.75 in.
Solids Syringe
25 G
2.75 in.
70 mm