Learn More
SGE™ SilFlow™ GC 3 Port Splitter Kits
Highly efficient and reliable micro fluidic platform to improve GC connectivity.
Brand:  SGE™ 123721
 View more versions of this product
Product Code. 12700948
  • 774.00 € / Each
Estimated Shipment: 04-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Kit includes a SilFlow microchannel device (MCD), SilTite FingerTite tool, mounting bracket, plus appropriate ferrules, nuts and blanking ferrules for set up
  • 1.1mm OD tubing not included
Specifications
Specifications
Port Splitter
0.53 mm
SilFlow microchannel device (MCD), SilTite FingerTite tool, Mounting bracket, Appropriately sized ferrules, nuts and blanking ferrules for set up
SGE Silflow Microchannel Device