Learn More
SGSH Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 28150-1-AP-20UL
 View more versions of this product
Product Code. 16886015
  • 145.00 € / 20µL
Explore more special offers
Description
Description
The SGSH antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SGSH. This antibody recognizes human antigen. The SGSH antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
SGSH
Polyclonal
Unconjugated
20 μL
6448
IgG
Western Blot, Immunohistochemistry, ELISA
300 μg/mL