Learn More
SGTA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11019-2-AP-150UL
 View more versions of this product
Product Code. 16837803
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.
Specifications
Specifications
SGTA
Polyclonal
Unconjugated
SGTA
Alpha SGT, alphaSGT, aSGT, hSGT, SGT, SGT1, SGTA, UBP, Vpu binding protein
Rabbit
Antigen Affinity Chromatography
RUO
52551, 6449, 64667
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43765, O70593, Q8BJU0
Sgta
SGTA Fusion Protein Ag1475
150 μL
Primary
Human, Mouse, Rat