missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SGTA Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11019-2-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein which is capable of interacting with the major nonstructural protein of parvovirus H-1 and 70-kDa heat shock cognate protein; however, its function is not known. Since this transcript is expressed ubiquitously in various tissues, this protein may serve a housekeeping function.Specifications
SGTA | |
Polyclonal | |
Unconjugated | |
SGTA | |
Alpha SGT, alphaSGT, aSGT, hSGT, SGT, SGT1, SGTA, UBP, Vpu binding protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
52551, 6449, 64667 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43765, O70593, Q8BJU0 | |
Sgta | |
SGTA Fusion Protein Ag1475 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |