Learn More
Manutan Shackles, 3.75T large D, high tensile steel
Product Code. 12801869
Encompass_Preferred
Product Code. 12801869
Brand: Manutan A340002
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use to securely connect lifting device to the load to be lifted.
  • Large Dee - Bolt
  • High tensile steel
  • Dimensions : d - 25mm ; D - 29mm ; w - 51mm ; s - 95mm ; e - 56mm