Learn More
Epredia™ Shandon™ Biopsy Processing/Embedding Cassettes II in Tube Packs, aqua
Shop All Epredia Products
Click to view available options
Color:
Aqua
Blue
Fluorescent Green
Fluorescent Pink
Fluorescent Yellow
Gray
Green
Lilac
Orange
Peach
Pink
Tan
White
Yellow
missing translation for 'unitSize'
Each
Pack of 750
Description
- Writing face is flat 45° angle
- Locking lid attached by two hinges
- Front or back opening
Ordering Information:
- Packaged in 75-cassette tubes
Specifications
Specifications
| Pore Style | Small Square Pore |
| Writing Face Angle | 45° |
| Color | Fluorescent Yellow |
| Product Type | Biopsy Cassette |