missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Bluing Reagent

Employ a gentle, pH-controlled solution for bluing hematoxylin with the Epredia™ Shandon™ Bluing Reagent.

57.40 € - 114.00 €

Specifications

Type Bluing Reagent
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10381775
View Documents Promotion Details
Epredia™
6769001
1L
57.40 €
Each
Please to purchase this item. Need a web account? Register with us today!
10127998
View Documents Promotion Details
Epredia™
6769002
4L
114.00 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Ready-to-use
  • Thoroughly pH-controlled for completely effective bluing
Specifications

Specifications

Bluing Reagent
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H370-Causes damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P235-Keep cool.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P363-Wash contaminated clothing before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium bicarbonate,lithium carbonate,methyl alcohol

SDS
Documents

Documents

Product Certifications