Learn More
Epredia™ Cryochrome™ Embedding Resin
Support tissue for frozen sectioning with Epredia™ Cryochrome™ Embedding Resin.
85.70 € - 130.56 €
Specifications
Product Type Embedding Media
Quantity 120mL
View More Specs
Products 6
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12533527
View Documents
Epredia™
9990422
Embedding Media This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12560017
View Documents
Epredia™
9990423
Embedding Media This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12726087
View Documents
Epredia™
9990426
Embedding Media This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12570017
View Documents
Epredia™
9990426
Embedding Media
106.00 €
Pack of 4
In Stock
to see stock.
Add to basket
12543527
View Documents
Epredia™
9990428
Embedding Media This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12580017
View Documents
Epredia™
9990429
Embedding Media This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Color codes frozen sections for easy identification and tracking
  • Four colors available: pink, blue, yellow, green
  • Formulated with physical characteristics identical to Cryomatrix™ embedding resin
Specifications
Specifications
Embedding Media
120mL