Learn More
Epredia™ Cryomatrix™ embedding resin
Secure and support tissue with Epredia™ Cryomatrix™ embedding resin, an outstanding product used to support tissue for frozen sectioning.
Brand: Epredia™ 6769006
36.10 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.
This item is not returnable.
View return policy
Description
- Forms a firm bond with the specimen holder
- Freezes quickly, sections smoothly
- Dissolves in water without residue
Specifications
120 mL | |
Embedding Media |