Learn More
Epredia™ Cytoclip™ Stainless-Steel Slide Clip
The Epredia™ Cytoclip Stainless-Steel Slide Clip holds Cytofunnel™ disposable sample chamber against the microscope slide.
Brand: Epredia™ 59910052
771.20 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- For preparation in the Epredia Shandon Cytospin™ Cytocentrifuge
- Autoclavable and reusable
Specifications
Stainless steel slide clip | |
6/Box |