Learn More
Epredia™ Double Cytofunnel™
Optimize cell yield with the Epredia™ Double Cytofunnel™.
Brand:  Epredia™ 5991039
170.80 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Product Code. 12016679
  • 244.00 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Produces more cost-effective, thin-layer preparations when compared to other thin-layer methods
  • Reduces the risk of exposure to pathogenic samples
  • Reduces preparation time
  • Disposable to eliminate time-consuming decontamination and cleaning
  • Epredia™ Double Cytofunnel™ REQUIRES Epredia™ Cytoclips™
  • Decreases the possibility of cross-contamination
  • Features funnel markings corresponding to slide markings for simple identification
Includes:
  • Pre-attached filter card
  • Closure caps
  • Easy-to-use operational illustrations
Specifications
Specifications
White
25 Pk.
0.05 to 0.25 mL
Single-use
Double Cytofunnel™, With Filter Cards and Caps