Learn More
Epredia™ Double Cytoslide™ Microscope Slides
Product Code. p-4525365 Shop All Epredia Products
Click to view available options
Description:
Coated
Noncoated
Unit Size:
Pack of 100
Product Code. 12006689
Brand: Epredia™ 5991054
Only null left
Add to basket
Add to basket
Use Epredia™ Shandon™ Double Cytoslides™ with Epredia™ Shandon™ EZ Double Cytofunnel™ Chambers.
  • Specimen is displayed within a circle marked as 1 or 2
  • Number circles correspond with the Cytofunnel™ chamber
Ordering Alert:
  • Sold in boxes of 100 slides
TRUSTED_SUSTAINABILITY
Specifications
Type Double Cytoslide
Color Plain
Description Noncoated