Learn More
Epredia™ Shandon™ EA-65 Stain
Improve the expression of eosin-stained components with Epredia™ Shandon™ EA-65 Stain, which contains a lower concentration of light green.
Brand:  Epredia™ 6766004
 View more versions of this product
Product Code. 12647936
  • 144.00 € / Each
Explore more special offers
Description
Description
  • Polychromatic cytoplasmic stain
  • Ideal for thicker non-GYN preparations
  • Biological Stain Commission-certified dye
Specifications
Specifications
Cellular Components Stain
4L