Learn More
Epredia™ Embedding Cassettes in Tube Packs
Hold and identify tissue samples in processing, embedding and sectioning procedures with Embedding Cassettes in Tube Packs. Compatible for use with the Epredia™ PrintMate AS™ Cassette Printer.
Brand: Epredia™ B1000732WH
This item is not returnable.
View return policy
Description
- Can be used on all Epredia PrintMate automatic cassette labelers
- Fit securely in microtome chuck adapters
- Designed to accept metal lids (Order number 1001003, 25/pk., sold separately)
- Made of acetal polymer with a pore style of round holes
- Supplied in 10 tubes of 75 cassettes each
Specifications
Round Holes | |
750 Cassettes/Cs. | |
White | |
Acetal Polymer |