Learn More
Epredia™ Shandon™ Embedding Rings
Identify samples in storage with Epredia™ Shandon™ Embedding Rings, which securely hold tissue samples for sectioning.
Brand: Epredia™ 1000956
75.68 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
This item is not returnable.
View return policy
Description
- Made of high impact polystyrene
- Fits well in microtome chuck adapters
- Supplied in boxes of 250 rings
Specifications
Blue | |
Polystyrene | |
Embedding Rings |