Learn More
Epredia™ Shandon™ Eosin-Y Stain
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
Brand:  Epredia™ 6766008
113.88 EUR valid until 2024-12-31
Use promo code "21984" to get your promotional price.
 View more versions of this product
Product Code. 10483750
  • 181.00 € / 4 L
Explore more special offers
Description
Description
  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations
Specifications
Specifications
Alcoholic Eosin Y Stain
4L
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Shandon Eosin-Y Alcoholic
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.