missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Eosin-Y Stain

Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.

Brand:  Epredia™ 6766007

38.44 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.



 View more versions of this product

Product Code. 10562614

  • 52.15 € / 2 L

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Ready-to-use
  • Produces consistent results
  • Stable and precipitate-free; no filtering
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic or aqueous formulations
Specifications

Specifications

Alcoholic Eosin Y Stain
1L
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 3
  • Germ cell mutagenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H350-May cause cancer.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.

Supplemental information
  • MIXTURE LIST-Contains : ethyl, alcohol, methyl alcohol, isopropyl alcohol, acetic acid, eosin-Y dye

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.