Learn More
Epredia™ Shandon™ Eosin-Y Stain
Produce brilliant staining with Epredia™ Shandon™ Eosin-Y, a fast-acting cytoplasmic counterstain.
Brand: Epredia™ 6766007
This item is not returnable.
View return policy
Description
- Ready-to-use
- Produces consistent results
- Stable and precipitate-free; no filtering
- Compatible with all alum hematoxylin stains
- Available in alcoholic or aqueous formulations
Specifications
Alcoholic Eosin Y Stain | |
1L |