missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Gill™ Hematoxylin

Product Code. p-4530263 Shop All Epredia Products
Click to view available options
Quantity:
4L
2 x 1L
Type:
Shandon Gill 1 Hematoxylin
Shandon Gill 2 Hematoxylin
Shandon Gill 3 Hematoxylin
Unit Size:
4 L
Each
Pack of 2
This item is not returnable. View return policy

Product Code. 12657926

Brand: Epredia™ 6765005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Achieve fast and convenient results, while maintaining complete control over stain intensity with Epredia™ Shandon™ Gill™ Hematoxylin, which has mucoid staining.

  • Gill 1: Light blue nuclei

  • Gill 2: Medium blue nuclei

  • Gill 3: Blue-black nuclei

TRUSTED_SUSTAINABILITY

Specifications

Type Shandon Gill 1 Hematoxylin
Quantity 2 x 1L
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • H371-May cause damage to organs.
Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contains : ethylene glycol,acetic acid, aluminium sulphate, sodium iodate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.