Learn More
Epredia™ Glyo-Fixx™
Obtain superior results to neutral buffered formalin with Glyo-Fixx™, a noncarcinogenic, nonsensitizing, and environmentally friendly substitute.
81.80 € - 260.00 €
Specifications
Product Type Fixative
View More Specs
Products 4
Product Code Brand Product Type Quantity Price Quantity & Availability  
Product Code Brand Product Type Quantity Price Quantity & Availability  
10249908
View Documents
Epredia™
6764260
Fixative 5 gal.
208.00 €
Each
10602689
View Documents
Epredia™
6764262
Fixative 1 gal.
81.80 €
3.80 L
12500067
View Documents
Epredia™
9990924
Fixative 125 x 60mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12520067
View Documents
Epredia™
9990928
Fixative 125 x 120mL
260.00 €
Pack of 125
Description
Description
  • Produces distinct and crisp cellular morphology
  • Fixes most antigens without impairing immunoreactivity
  • Allows special stain dyes, including silver procedures, to be sharp without background staining
  • Presents fine detail in specimen structure
Specifications
Specifications
Fixative