missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Harris Hematoxylin

Take advantage of acidified or non-acidified formulations with the Epredia™ Shandon™ Harris Hematoxylin. This product does not employ a mercury-based oxidizing agent, thus removing a possible source of environmental contamination.

75.90 € - 160.00 €

Specifications

Type Hematoxylin
View More Specs

Products 4
Product Code Brand Type Quantity Price Quantity & Availability  
Product Code Brand Type Quantity Price Quantity & Availability  
12627926
View Documents Promotion Details
Epredia™
6765001
Hematoxylin 2 x 1L
75.90 €
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
12637926
View Documents Promotion Details
Epredia™
6765002
Hematoxylin 4L
160.00 €
Each
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
12647926
View Documents Promotion Details
Epredia™
6765003
Hematoxylin 2 x 1L
134.00 €
Pack of 2
Estimated Shipment: 20-02-2025
to see stock.
Add to basket
 
10694409
View Documents Promotion Details
Epredia™
6765004
Hematoxylin 4L
125.00 €
Each
Estimated Shipment: 18-11-2024
to see stock.
Add to basket
 
Description

Description

  • Acidifed: Intensely stained, blue-black nuclei

  • Non-acidified: Intensely stained, blue-black nuclei

Specifications

Specifications

Hematoxylin
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2

Hazard Statement
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H371-May cause damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P271-Use only outdoors or in a well-ventilated area.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium iodate, aluminium ammonium sulphate dodecahydrate, hematoxylin, ethyl alcohol, methyl alcohol

SDS
Documents

Documents

Product Certifications