missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Accessories for Hyperclean TruAir™ Workstation
For use with Epredia™ Hyperclean Pathology Workstations.
Brand: Epredia™ 9990612
139.52 EUR valid until 2025-02-28
Use promo code "24154" to get your promotional price.
This item is not returnable.
View return policy
Description
Accessories:- Charcoal Filter
- Fine particle Filters
- Formaldehyde Neutralizing (Potassium Permanganate) Filter
Specifications
Formaldehyde Neutralizing Filter | |
Size: 28.7 cm x 14.7 cm x 3.5 cm | |
Epredia Hyperclean Pathology Workstations | |
Hyperclean™ |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H371-May cause damage to organs.
Precautionary Statement
- P235-Keep cool.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P314-Get medical advice/attention if you feel unwell.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : aluminium oxide, potassium permanganate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction