missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Instant Eosin

Obtain brilliant and consistent staining with Epredia™ Shandon™ Instant Eosin.

Brand:  Epredia™ 6765540


 View more versions of this product

Product Code. 10046318

  • 238.00 € / Pack of 6
Estimated Shipment: 23-10-2024
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stable and precipitate-free; no filtering
  • pH-adjusted for maximum stain uptake
  • Economical powder stores indefinitely
  • Compatible with all alum Hematoxylin stains
Specifications

Specifications

Alum Hematoxylin Stains
Eosin
Liquid
Eosin
Healthcare
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P264-Wash face, hands, and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : monosodium phosphate, sodium phosphate dibasic, eosin-y dye,sucrose

SDS
Documents

Documents

Certificates
Special Offers

Special Offers