missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Instant Hematoxylin

Substitute Epredia™ Shandon™ Instant Hematoxylin for Harris or Gill when no mucoid staining is required.

Brand:  Epredia™ 6765015

158.20 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.



Product Code. 12687926

  • 232.00 € / Pack of 6
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Stains from blue-black to light blue depending upon preparation
Specifications

Specifications

Instant Hematoxylin
Makes 1L x 6; 6/pack
Makes 1L x 6; 6/pack
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Shandon Instant Hematoxylin

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.