Learn More
Epredia™ Shandon™ Instrument Case
Epredia™ Shandon Instrument Case is available as a complete kit or case only.
8715.00 €
Description
Case dimensions:
- 12.75 x 9.5 x 2.5 in. (32.4 x 24.1 x 6.4cm)
Instruments Available in Varying Sizes and Styles:
- Dissecting scissors
- Iris scissors
- Liston bone shears
- Rib shears
- Plain bone shears
- Liston bone-cutting forceps
- Councilman's chisel
- Tissue thumb forcep
- Broad point dressing thumb forceps
- Cartilage thumb forceps
- Halstead's hemostatic mosquito forceps
- Dura strip forceps
- Virchow skull breaker
- Bone mallet
- Post mortem hammer with hook
- Grooved needle director
Specifications
Instrument Case |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Epredia™ Shandon™ Instrument Case