missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Kwik-Diff™ Stains
Enjoy a quicker alternative to traditional Wright's and Giemsa Staining procedures with Epredia™ Shandon™ Kwik-Diff™ Stains.
70.00 € - 272.00 €
Product Code | Brand | Type | Includes | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Includes | Quantity | Price | Quantity & Availability | |||
10435310
|
Epredia™
9990700 |
Kwik-Diff Stains | Kit includes 500 mL bottle each: Fixative, Eosin, Methylene Blue | 3 x 500 mL |
272.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10633649
|
Epredia™
9990705 |
Fixative | (4 L) Kwik-Diff Reagent 1, Fixative | 4 L |
163.00 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10643649
|
Epredia™
9990706 |
Eosin | (4 L) Kwik-Diff Reagent 2, Eosin | 4 L |
156.00 €
Each |
In Stock
Log in to see stock. |
|||
10549408
|
Epredia™
9990707 |
Methylene Blue | (4 L) Kwik-Diff Reagent 3, Methylene Blue | 4 L |
155.00 €
Each |
In Stock
Log in to see stock. |
|||
12539956
|
Epredia™
9990701 |
Kwik-Diff Stains | Kit includes 125 mL bottle each: Fixative, Eosin, Methylene Blue | 125 mL | N/A | ||||
Description
- Available in kit form or as individual components
- Fast three-step differential stain for bone marrows, smears, FNAs, and microorganisms such as H. pylori
Reagent 1: Fixative; Reagent 2: Eosin; Reagent 3: Methylene Blue
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Shandon Kwik Diff Solution 1 - Methanol Fixative
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H370-Causes damage to organs.
- H371-May cause damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P235-Keep cool.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title