missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Neoprene Cork Dissecting Boards, Small

Epredia™ Shandon Neoprene Cork Dissecting Boards feature a durable cutting surface over a marine plywood base.

Brand:  Epredia™ 361

 View more versions of this product

Product Code. 15323575

This item is not returnable. View return policy

Description

Description

Features:

  • A 0.25 in. (6mm) layer of neoprene cork is permanently cemented to base.
  • Blended rubber and cork composition resists absorption and allows for thorough rinsing.
  • Rubber bumpers attached to each corner raise the cutting surface to 1.5 in. (3.8cm).

Ordering information:

  • Large board measures 16 x 23 x 0.75 in. (40.6 x 58.4 x 1.9cm)
  • Small board measures 12 x 16 x 0.75 in. (50.5 x 40.6 x 1.9cm)
  • Board fits Woll Drain Dissecting Tray 360
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dissecting Boards
Large
16 in.
23 in.
0.75 in.
Blended Rubber and Cork
16 x 23 x 0.75 in. (40.6 x 58.4 x 1.9 cm)
40.6 cm
58.4 cm
1.9 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Epredia™ Shandon™ Neoprene Cork Dissecting Boards, Small > Large board

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.