missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Shandon™ Nu-Clear™ Reagent

Achieve improved cell clarity by removing excess hematoxylin from the slide with Epredia™ Shandon™ Nu-Clear™, a specially-formulated acid alcohol solution.

78.95 €

Specifications

Product Type Tissue Processing Reagent
View More Specs

Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12513627
View Documents
Epredia™
6769009
Tissue Processing Reagent
78.95 €
4 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Key Features 
  • Removes residual hematoxylin staining the glass.
  • Designed to be used on the full line of Epredia Shandon automatic stainers as well as manual methods.

Nu-Clear I

  • Fast-acting solution
  • Recommended to be used as a 10-second step in most protocols
  • Particularly beneficial for use with Harris hematoxylin
  • May be used on the Epredia Varistain™ 24-4 or Shandon Varistain Gemini slide stainers

Nu-Clear II

  • Less concentrated
  • Recommended as a 20-second step
  • May be used on the Epredia Shandon Linistat and other linear stainers
  • Rcommended for use with Gill hematoxylins
Specifications

Specifications

Tissue Processing Reagent
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Carcinogenicity Category 1A
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H350-May cause cancer.
  • H370-Causes damage to organs.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P242-Use only non-sparking tools.
  • P243-Take precautionary measures against static discharge.
  • P235-Keep cool.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : ehtyl alcohol, methyl alcohol, methylisobutyl ketone, hydrochloric acid

SDS
Documents

Documents

Product Certifications