Learn More
Epredia™ Shandon™ TBD-2™ Decalcifier
Process specimens up to 48 hours past end point undamaged with Epredia™ Shandon™ TBD-2™ Decalcifier, a formic acid decalcifier.
168.00 €
Specifications
Product Type Decalcifying Solution
View More Specs
Products 1
Product Code Brand Product Type Price Quantity & Availability  
Product Code Brand Product Type Price Quantity & Availability  
12607926
View Documents
Epredia™
6764004
Decalcifying Solution
168.00 €
4 L
Description
Description
  • Protects cells from actions of acid with added ingredients
  • Permits utilization of routine end point tests, such as calcium oxalate
Specifications
Specifications
Decalcifying Solution
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Shandon TBD-2 Decalcifier
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P234-Keep only in original container.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P406-Store in corrosive resistant/ container with a resistant inner liner.