missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Shandon™ Tissue-Marking Dyes, Tissue marking dye kit
Permanently mark the margins of excised surgical specimens for exact orientation with Epredia™ Shandon™ Tissue-Marking Dyes.
31.05 € - 233.00 €
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Price | Quantity & Availability | ||||
10799839
|
Epredia™
3120123 |
Multiple | 5 x 59 mL |
233.00 €
Each |
In Stock
Log in to see stock. |
||||
10413229
|
Epredia™
3120124 |
Blue | 59 mL |
34.20 €
Each |
In Stock
Log in to see stock. |
||||
10830020
|
Epredia™
3120125 |
Black | 59 mL | N/A | |||||
10530159
|
Epredia™
3120126 |
Yellow | 59 mL |
31.05 €
Each |
In Stock
Log in to see stock. |
||||
10692479
|
Epredia™
3120127 |
Red | 59 mL |
35.90 €
Each |
In Stock
Log in to see stock. |
||||
10415339
|
Epredia™
3120128 |
Green | 59 mL |
38.95 €
Each |
In Stock
Log in to see stock. |
||||
Description
- Special binding pigment evenly coats the tissue without penetrating tissue surface
- Does not interfere with diagnostic interpretation
- Quick-drying and easy-to-use; decreases time-factor in urgent frozen-sectioning
Ordering Information:
Available individually or as a kit (includes one bottle each of black, blue, yellow, red, and green; 50 applicator sticks; storage base).
Specifications
Tissue Marking Dye |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H318-Causes serious eye damage.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- H371-May cause damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P202-Do not handle until all safety precautions have been read and understood.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : carbon black, ammonium hydroxide, formaldehyde
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title