missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Xylene Substitute Mountant
Employ all routine xylene-substitute clearing agents with the Epredia™ Xylene Substitute Mountant.
52.45 € - 174.00 €
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
13300125
|
Epredia™
1900233 |
120 mL |
52.45 €
120mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10056318
|
Epredia™
9999122 |
500 mL |
174.00 €
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Permanent, organic mounting medium
- Toluene base
- Refractive index: 1.495 ±0.005 (dry)
- Viscosity: Medium (120-265 cps at 24°C)
Specifications
Xylene Substitute |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Shandon Xylene Substitute Mountant
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H336-May cause drowsiness or dizziness.
- H361d-Suspected of damaging the unborn child.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- EUH208-Contains . May produce an allergic reaction.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P370+P378-In case of fire: Use CO2, dry chemical, or foam to extinguish.
- P391-Collect spillage.
- P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
- MIXTURE LIST-Contains : toluene, isobutyl methacrylate
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title