missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Xylene Substitute
Achieve safer, more convenient processing, staining and coverslipping with Epredia™ Xylene Substitute.
125.63 € - 289.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10524305
|
Epredia™
6764506 |
4 x 1 gal. |
289.00 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10128638
|
Epredia™
9990505 |
1 gal. |
125.63 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Not sensitizing to normal skin
- Evaporation rate similar to xylene
- Virtually odorless aliphatic hydrocarbon—no citrus aroma
- Airborne exposure limit three times safer than xylene (TLV 300ppm)
Specifications
Xylene Substitute |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Carcinogenicity Category 1B
- Germ cell mutagenicity Category 1B
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H341-Suspected of causing genetic defects.
- H351-Suspected of causing cancer.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P262-Do not get in eyes, on skin, or on clothing.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
- MIXTURE LIST-Contains : naptha (petroleum),hydrotreated heavy