Learn More
SharpSafe™ Polypropylene Containers
Use yellow SharpSafe™ Polypropylene Containers for the collection of sharp waste treatment activities and instruments that are pungent, sharp and/or pointed.
Specifications
Certifications/Compliance | Complies with UN 3291 and BS 7320 standard, NF-X-30–500, NF302, A.D.R. and UN3291, TRBA 250, Quality management system: ISO 9001:2008 and ISO 13485:2003, Environmental Standard: ISO 14001:2004 |
---|---|
Closure Type | Flip Top |
Closure Color | Translucent |
Color | Yellow |
Autoclavable | Autoclavable |
Description
- Translucent flip-top lid and handle
- Yellow
- Polypropylene
- Autoclavable
Specifications
Complies with UN 3291 and BS 7320 standard, NF-X-30–500, NF302, A.D.R. and UN3291, TRBA 250, Quality management system: ISO 9001:2008 and ISO 13485:2003, Environmental Standard: ISO 14001:2004 | |
Translucent | |
Autoclavable | |
Polypropylene |
Flip Top | |
Yellow | |
Flip top lid, handle |