Learn More
Sharpsafe™ Polypropylene Medicinal Sharps Container
Designed for use in public access areas such as A&E and Out-patients. Sharpsafe™ Polypropylene Medicinal Sharps Container allows easy disposal of sharps but provides added security for both the user and the general public. The Protected Access range also discourages the use of the sharps container for non sharps waste, avoiding unnecessary waste disposal costs.
Brand:  Sharpsafe™ 41105430
 View more versions of this product
Product Code. 12903535
Description
Description
  • Easy disposal of sharps
  • Protected Access range 
  • Polypropylene
Specifications
Specifications
7 L
Lid
Yellow
1 Ea.
Designed for use in public access areas such as A&E and out-patients
BS 7320, AFNOR NX30-500, UN 3291
Yellow
4110
Closure
Polypropylene
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Sharpsafe™ Polypropylene Medicinal Sharps Container > Capacity: 7L; Color: yellow